Assay Details
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046387.1 | NP_001039852.1 | 1 - 2 | 161 | 119 | |
GenBank mRNA | BC111154.1 | - | 1 - 2 | 161 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
