Assay Details
Gene Symbol: BNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046539.5 | NP_001040004.2 | 1 - 2 | 91 | 96 | |
GenBank mRNA | BC110132.1 | - | 1 - 2 | 95 | 96 | |
EE229689.1 | - | 1 - 2 | 92 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110132.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
