Assay Details
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046551.2 | NP_001040016.1 | 2 - 3 | 244 | 85 | |
GenBank mRNA | AM086628.1 | - | 2 - 3 | 154 | 85 | |
BC112547.1 | - | 2 - 3 | 245 | 85 | ||
EU276062.1 | - | 2 - 3 | 192 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
