Assay Details
Gene Symbol: NARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046572.2 | NP_001040037.1 | 8 - 9 | 687 | 98 | |
GenBank mRNA | BC105358.1 | - | 8 - 9 | 688 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
