Assay Details
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075166.1 | NP_001068634.1 | 5 - 6 | 779 | 68 | |
GenBank mRNA | BC109588.1 | - | 5 - 6 | 779 | 68 | |
EU005652.1 | - | 5 - 6 | 802 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
