Assay Details
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075797.3 | NP_001069265.2 | 4 - 5 | 487 | 150 | |
GenBank mRNA | BC109850.1 | - | 4 - 5 | 486 | 150 | |
EV618408.1 | - | 4 - 5 | 488 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
