Assay Details
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076301.2 | NP_001069769.1 | 1 - 2 | 68 | 80 | |
GenBank mRNA | BC109863.1 | - | 1 - 2 | 69 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109863.1, NM_001076301.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
