Assay Details
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076366.1 | NP_001069834.1 | 363 | 112 | ||
GenBank mRNA | BC110012.1 | - | 363 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
