Assay Details
Gene Symbol: GPR161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077066.1 | NP_001070534.1 | 4 - 5 | 1287 | 111 | |
GenBank mRNA | BC110145.1 | - | 4 - 5 | 1287 | 111 | |
BT026556.1 | - | 4 - 5 | 1290 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
