Assay Details
Gene Symbol: GPR161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077066.1 | NP_001070534.1 | 5 - 6 | 1413 | 88 | |
GenBank mRNA | BC110145.1 | - | 5 - 6 | 1413 | 88 | |
BT026556.1 | - | 5 - 6 | 1416 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
