Assay Details
Gene Symbol: DUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077873.2 | NP_001071341.1 | 502 | 103 | ||
GenBank mRNA | BC111641.1 | - | 503 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
