Assay Details
Gene Symbol: ABHD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015606.2 | NP_001015606.1 | 1 - 2 | 160 | 91 | |
GenBank mRNA | BC112739.1 | - | 1 - 2 | 160 | 91 | |
BT020941.1 | - | 1 - 2 | 148 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
