Assay Details
Gene Symbol: MRPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045996.2 | NP_001039461.1 | 3 - 4 | 468 | 79 | |
GenBank mRNA | BC105311.1 | - | 3 - 4 | 469 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
