Assay Details
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046068.2 | NP_001039533.1 | 5 - 6 | 675 | 78 | |
GenBank mRNA | BC105368.1 | - | 5 - 6 | 676 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
