Assay Details
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046069.2 | NP_001039534.1 | 2 - 3 | 322 | 87 | |
GenBank mRNA | BC112484.1 | - | 2 - 3 | 323 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
