Assay Details
Gene Symbol: NDUFAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046105.2 | NP_001039570.1 | 3 - 4 | 362 | 125 | |
GenBank mRNA | BC105350.1 | - | 3 - 4 | 363 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
