Assay Details
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046173.1 | NP_001039638.1 | 9 - 10 | 1060 | 70 | |
GenBank mRNA | BC105529.1 | - | 9 - 10 | 1060 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
