Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046204.2 | NP_001039669.2 | 22 - 23 | 2770 | 66 | |
GenBank mRNA | BC114145.1 | - | 8 - 9 | 1014 | 66 | |
BC148971.1 | - | 22 - 23 | 2634 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
