Assay Details
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046397.2 | NP_001039862.1 | 1 - 2 | 127 | 148 | |
GenBank mRNA | BC112681.1 | - | 1 - 2 | 128 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
