Assay Details
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075179.1 | NP_001068647.1 | 5 - 6 | 517 | 135 | |
GenBank mRNA | BC118438.1 | - | 5 - 6 | 517 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
