Assay Details
Gene Symbol: POLR3GL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075213.1 | NP_001068681.1 | 1 - 2 | 68 | 91 | |
GenBank mRNA | BC114765.1 | - | 1 - 2 | 68 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114765.1, NM_001075213.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
