Assay Details
Gene Symbol: ARPC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075359.1 | NP_001068827.1 | 3 - 4 | 274 | 82 | |
GenBank mRNA | BC147946.1 | - | 3 - 4 | 343 | 82 | |
BT025475.1 | - | 3 - 4 | 274 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
