Assay Details
Gene Symbol: ANGPTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075426.1 | NP_001068894.1 | 1 - 2 | 299 | 77 | |
GenBank mRNA | BC114763.1 | - | 1 - 2 | 299 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114763.1, NM_001075426.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
