Assay Details
Gene Symbol: UBE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075711.1 | NP_001069179.1 | 3 - 4 | 405 | 80 | |
GenBank mRNA | BC116118.1 | - | 3 - 4 | 405 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
