Assay Details
Gene Symbol: DNASE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075865.1 | NP_001069333.1 | 3 - 4 | 414 | 124 | |
GenBank mRNA | BC118435.1 | - | 3 - 4 | 414 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
