Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075902.2 | NP_001069370.2 | 6 - 7 | 489 | 70 | |
GenBank mRNA | BC114844.1 | - | 6 - 7 | 490 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
