Assay Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075960.1 | NP_001069428.1 | 4 - 5 | 533 | 95 | |
GenBank mRNA | BC116148.1 | - | 4 - 5 | 533 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
