Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076031.1 | NP_001069499.1 | 4 - 5 | 491 | 76 | |
GenBank mRNA | BC116135.1 | - | 4 - 5 | 491 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
