Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076121.2 | NP_001069589.1 | 4 - 5 | 998 | 101 | |
GenBank mRNA | BC118227.1 | - | 4 - 5 | 856 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
