Assay Details
Gene Symbol: KDELR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076495.1 | NP_001069963.1 | 1 - 2 | 237 | 70 | |
GenBank mRNA | BC118250.1 | - | 1 - 2 | 237 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
