Assay Details
Gene Symbol: OGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077019.1 | NP_001070487.1 | 2 - 3 | 225 | 70 | |
GenBank mRNA | BC105559.1 | - | 2 - 3 | 225 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
