Assay Details
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077105.2 | NP_001070573.1 | 3 - 4 | 546 | 91 | |
GenBank mRNA | AB689695.1 | - | 3 - 4 | 513 | 91 | |
BC114846.1 | - | 3 - 4 | 547 | 91 | ||
LC271182.1 | - | 3 - 4 | 485 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
