Assay Details
Gene Symbol: AP1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077110.1 | NP_001070578.1 | 3 - 4 | 935 | 74 | |
GenBank mRNA | BC116132.1 | - | 3 - 4 | 935 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
