Assay Details
Gene Symbol: FGF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079789.1 | NP_001073257.1 | 4 - 5 | 405 | 116 | |
GenBank mRNA | BC118169.1 | - | 4 - 5 | 405 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
