Assay Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174702.3 | NP_777127.1 | 2 - 3 | 574 | 64 | |
GenBank mRNA | AF028005.1 | - | 2 - 3 | 568 | 64 | |
AH004629.2 | - | 2 - 3 | 590 | 64 | ||
BC105525.1 | - | 2 - 3 | 574 | 64 | ||
BT025412.1 | - | 2 - 3 | 600 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
