Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182786.2 | NP_877587.1 | 1 - 2 | 284 | 90 | |
GenBank mRNA | AY322482.1 | - | 1 - 2 | 279 | 90 | |
BC118280.1 | - | 1 - 2 | 284 | 90 | ||
BT029837.1 | - | 1 - 2 | 285 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
