Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182786.2 | NP_877587.1 | 2 - 3 | 538 | 77 | |
GenBank mRNA | AY322482.1 | - | 2 - 3 | 533 | 77 | |
BC118280.1 | - | 2 - 3 | 538 | 77 | ||
BT029837.1 | - | 2 - 3 | 539 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
