Assay Details
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205813.2 | NP_991382.2 | 8 - 9 | 1322 | 95 | |
GenBank mRNA | AY344127.1 | - | 9 - 10 | 1198 | 95 | |
BC113344.1 | - | 8 - 9 | 1322 | 95 | ||
BT030591.2 | - | 8 - 9 | 1164 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
