Assay Details
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038198.2 | NP_001033287.1 | 4 - 5 | 1209 | 60 | |
GenBank mRNA | AY644769.1 | - | 2 - 3 | 990 | 60 | |
AY644770.1 | - | 2 - 3 | 993 | 60 | ||
BC123390.1 | - | 4 - 5 | 1209 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
