Assay Details
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075152.1 | NP_001068620.1 | 1 - 2 | 257 | 134 | |
GenBank mRNA | BC119932.1 | - | 1 - 2 | 257 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC119932.1, NM_001075152.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
