Assay Details
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075246.2 | NP_001068714.1 | 8 - 9 | 993 | 90 | |
GenBank mRNA | BC120275.1 | - | 8 - 9 | 995 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
