Assay Details
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075281.2 | NP_001068749.1 | 2 - 3 | 357 | 91 | |
GenBank mRNA | BC120197.1 | - | 2 - 3 | 358 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
