Assay Details
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075281.2 | NP_001068749.1 | 4 - 5 | 612 | 109 | |
GenBank mRNA | BC120197.1 | - | 4 - 5 | 613 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
