Assay Details
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075370.1 | NP_001068838.1 | 5 - 6 | 494 | 124 | |
GenBank mRNA | BC120067.1 | - | 5 - 6 | 494 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
