Assay Details
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075378.1 | NP_001068846.1 | 3 - 4 | 732 | 135 | |
GenBank mRNA | BC120317.1 | - | 3 - 4 | 732 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
