Assay Details
Gene Symbol: BCL2L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075417.2 | NP_001068885.1 | 1 - 2 | 334 | 76 | |
GenBank mRNA | BC122810.1 | - | 1 - 2 | 335 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
