Assay Details
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075445.1 | NP_001068913.1 | 2 - 3 | 327 | 98 | |
GenBank mRNA | BC120390.1 | - | 2 - 3 | 327 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
