Assay Details
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075559.2 | NP_001069027.1 | 2 - 3 | 250 | 90 | |
GenBank mRNA | BC123552.1 | - | 2 - 3 | 251 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
