Assay Details
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075730.1 | NP_001069198.1 | 8 - 9 | 1096 | 88 | |
GenBank mRNA | BC142342.1 | - | 9 - 10 | 881 | 88 | |
BT026272.1 | - | 8 - 9 | 1096 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
