Assay Details
Gene Symbol: AIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075813.2 | NP_001069281.1 | 2 - 3 | 418 | 85 | |
GenBank mRNA | BC122837.1 | - | 2 - 3 | 419 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
